Order Entry
Poland
Orders LinkContactUsLinkComponent
Benzaldehyde, pure

Benzaldehyde, pure

Glass bottle, 1 L
Catalog # CHMP421615306.1000
Irritant
Supplier:   CHEMPUR
Benzaldehyde, pure
Catalog # CHMP421615306.1000
Supplier:  CHEMPUR
CAS Number:  100-52-7
Benzaldehyde, pure
Benzaldehyde, pure

Glass bottle, 1 L

CAS Number:  100-52-7
Price
285 zł
Log in for availability
1 * 1 L
Product Details & Documents
  • CAS Number:100-52-7
  • Formula:C₆H₅CHO
  • MDL Number:MFCD00003299
  • Molecular Weight:106.12 g/mol
  • Merck Index:13,01057
  • Boiling Pt:179 °C (1013 hPa)
  • Melting Pt:–26 °C
  • Density:1.049 g/cm³ (20 °C)
  • Flash Pt:64 °C
  • Storage Temperature:Ambient
  • EINECS:202-860-4
  • UN:1990
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications