Order Entry
Poland
ContactUsLinkComponent
Recombinant protein, kiaa0513( aah30280.1, 1 a.a. - 301 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009764-P01
Supplier:  Abnova
Recombinant protein, kiaa0513( aah30280.1, 1 a.a. - 301 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009764-P01
Supplier:  Abnova