0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
Recombinant protein, mycl1( np_005367.1, 1 a.a. - 206 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
ABNOH00004610-P01
:
Abnova
Recombinant protein, mycl1( np_005367.1, 1 a.a. - 206 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
ABNOH00004610-P01
:
Abnova
sds_ABNOH00004610-P01.pdf
TDS_ABNOH00004610-P01.pdf
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
: Merck
VWR® Facial Tissues
: VWR Collection