Order Entry
Poland
Orders LinkContactUsLinkComponent
Zinc, pellets

Zinc, pellets

Glass bottle for solids, 500 g
Catalog # CHMP422631359.0500
Supplier:   CHEMPUR
Zinc, pellets
Catalog # CHMP422631359.0500
Supplier:  CHEMPUR
CAS Number:  7440-66-6
Zinc, pellets

Glass bottle for solids, 500 g

CAS Number:  7440-66-6
Price
1 260 zł
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:7440-66-6
  • Formula:Zn
  • MDL Number:MFCD00011291
  • Molecular Weight:65.39 g/mol
  • Merck Index:12,10255
  • Boiling Pt:908 °C (1013 hPa)
  • Melting Pt:420 °C
  • Density:7.14 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-175-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications