Order Entry
Poland
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid, pure analytical reagent

L(+)-Ascorbic acid, pure analytical reagent

Glass bottle for solids, 1 kg
Catalog # CHMP115291509.1000
Supplier:   CHEMPUR
L(+)-Ascorbic acid, pure analytical reagent
Catalog # CHMP115291509.1000
Supplier:  CHEMPUR
CAS Number:  50-81-7
L(+)-Ascorbic acid, pure analytical reagent
L(+)-Ascorbic acid, pure analytical reagent

Glass bottle for solids, 1 kg

CAS Number:  50-81-7
Price
208 zł
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…192 °C
  • Density:1.694 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications