Order Entry
Poland
ContactUsLinkComponent
44 COMPOSES A 2MG/L CHAQUE DANS METHANOL (AMPOULES DE 1 ML ) Benzene [CAS:71-43-2] Chlorobenzene [CAS:108-90-7] 1 ,2-Dichlorobenzene [CAS:95-50-1] 1,3-Dichlorobenzene [CAS:541-73-1] 1,4-Dichlorobenzene [CAS:106-46-7] Ethylbenzene [CAS:100 M 1 * 10 Ampoul
Catalog # CMPL6216.2K.ML
44 COMPOSES A 2MG/L CHAQUE DANS METHANOL (AMPOULES DE 1 ML ) Benzene [CAS:71-43-2] Chlorobenzene [CAS:108-90-7] 1 ,2-Dichlorobenzene [CAS:95-50-1] 1,3-Dichlorobenzene [CAS:541-73-1] 1,4-Dichlorobenzene [CAS:106-46-7] Ethylbenzene [CAS:100 M 1 * 10 Ampoul
Catalog # CMPL6216.2K.ML
Frequently Bought Together