Order Entry
Poland
Orders LinkContactUsLinkComponent
Stators, Rheodyne® | Avantor
Stators, Rheodyne®
7980-004 DuraLife® II stator for MXP7980-000
numer katalogowy 554-2664
Dostawca:   IDEX RHEODYNE
Stators, Rheodyne®
numer katalogowy 554-2664
Dostawca:  IDEX RHEODYNE
Stators, Rheodyne®
Stators, Rheodyne®
7980-004 DuraLife® II stator for MXP7980-000
Przejdź do:
Price
Zapytaj o cenę
Zaloguj się dla dostępności
1 * 1 SZT
Product Details & Documents
Stators are available in 316 stainless steel, PEEK and proprietary materials. Rheodyne® stator materials have been researched and selected for their physical and mechanical strengths. Stators only need replacing if the ports or sealing surfaces become damaged.
DuraLife® stators are stainless steel stators with the wear surface coated to prolong valve life.
DuraLife® II stators are titanium base metal with coated wear surfaces.
Dane techniczne
  • Description:
    7980-004 DuraLife® II stator for MXP7980-000
  • Seq:
    01