Order Entry
Poland
Orders LinkContactUsLinkComponent
Cuvettes for spectrophotometers

Cuvettes for spectrophotometers

Macro, 7 ml, 20 mm, Quartz
Catalog # JENW83070-80
Supplier:   Cole-Parmer
Cuvettes for spectrophotometers
Catalog # JENW83070-80
Supplier:  Cole-Parmer
Cuvettes for spectrophotometers
Cuvettes for spectrophotometers

Macro, 7 ml, 20 mm, Quartz

Price
1 680 zł
Log in for availability
1 * 1 SZT
Product Details & Documents

Extensive range of cuvettes for spectrophotometry for measuring liquids.

Glass cuvettes are designed for the visible region whereas quartz cuvettes may be used in both the UV and visible regions.

Specifications
  • Path length:
    20 mm
  • Type:
    Macro
  • Volume:
    7 ml
  • Window material:
    Quartz
  • Seq:
    006
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.