Order Entry
Poland
Orders LinkContactUsLinkComponent
GC septa
GC septa
HT septa for optimised bleed and temperature, combined with outstanding mechanical properties, 11,5 mm, 400 °C
Catalog # 568-0363
GC septa
Catalog # 568-0363
GC septa
GC septa
HT septa for optimised bleed and temperature, combined with outstanding mechanical properties, 11,5 mm, 400 °C
Price
318 zł
Log in for availability
1 * 25 SZT
Product Details & Documents
Specifications
  • Ø:
    11,5 mm
  • Max. temp.:
    400 °C
  • Description:
    HT septa for optimised bleed and temperature, combined with outstanding mechanical properties
  • Seq:
    13