Ribbons for IP™ label printing system
IP-R-6407, 6400 ribbon series resin formulation, width 110 mm, 300 m, Black
BRDY110381
: Brady
These IP® printer ribbons are high density ribbon with excellent solvent resistance, good heat resistance and super scratch resistance and have ribbon spindle.
- Colour:Black
- Description:IP-R-6407, 6400 ribbon series resin formulation, width 110 mm
- Length:300 m
- Seq:14