Order Entry
Poland
Orders LinkContactUsLinkComponent
Base materials for microbiology media

Base materials for microbiology media

Proteose peptone, 500 g
Catalog # HIMERM005-500G
Base materials for microbiology media
Catalog # HIMERM005-500G
Supplier:  
CAS Number:  
Base materials for microbiology media

Proteose peptone, 500 g

CAS Number:  
Discontinued
Product Details & Documents
Specifications

Specifications

  • Pk:
    500 g
  • Description:
    Proteose peptone
  • Cat. No.:
    HIMERM005-500G
  • Storage temperature:
    15…25 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.