Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Nickel(II) carbonate basic hydrate, Sigma-Aldrich®

Glass bottle for solids, 250 g
Catalog # SIAL72248-250G
Supplier:   Merck
Nickel(II) carbonate basic hydrate, Sigma-Aldrich®
Catalog # SIAL72248-250G
Supplier:  Merck
CAS Number:  12607-70-4
Nickel(II) carbonate basic hydrate, Sigma-Aldrich®

Glass bottle for solids, 250 g

CAS Number:  12607-70-4
Price
291 zł
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Nickel(II) carbonate hydroxide hydrate, Basic nickel carbonate
  • CAS Number:12607-70-4
  • Formula:~NiCO₃ · 2Ni(OH)₂ · 4H₂O
  • MDL Number:MFCD00016262
  • Molecular Weight:304.11 g/mol (anhy)
  • EINECS:235-715-9

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications