Order Entry
Poland
ContactUsLinkComponent
HYDROCHL. ACID SOL.,6 M IN H2O, F. AMINOACID ANAL. 1 * 2 mL
Catalog # SIAL84429-10X2ML
Supplier:  Merck
HYDROCHL. ACID SOL.,6 M IN H2O, F. AMINOACID ANAL. 1 * 2 mL
Catalog # SIAL84429-10X2ML
Supplier:  Merck
Frequently Bought Together