Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Copper(I) oxide, Sigma-Aldrich®

5 g
Catalog # SIAL566284-5G
Irritant
Environment
Supplier:   Merck
Copper(I) oxide, Sigma-Aldrich®
Catalog # SIAL566284-5G
Supplier:  Merck
CAS Number:  1317-39-1
Copper(I) oxide, Sigma-Aldrich®

5 g

CAS Number:  1317-39-1
Price
372 zł
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Cuprous oxide, Dicopper oxide
  • CAS Number:1317-39-1
  • Formula:Cu₂O
  • MDL Number:MFCD00010974
  • Molecular Weight:143.09 g/mol
  • Merck Index:13,02694
  • Boiling Pt:1800 °C (1013 hPa)
  • Melting Pt:1232 °C
  • Density:5.7…6.2 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-270-7
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.