D-LACTOSE MONOHYDRATE BIOULTRA, >= 99.5% (HPLC) 1 * 25 g
Catalog # SIAL61339-25G
Supplier: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Citric acid monohydrate ≥98%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Reagents for Karl Fischer titrations, Aquastar®, Supelco®
Supplier: Merck
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Beakers, Low Form
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Orthophosphoric acid 85% ≥85%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Laboratory glass cleaner, borer 21 LAB CLF
Supplier: Borer Chemie
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
1,6-ANHYDRO-BETA-D-GLUCOSE, 99% 1 * 1 g
Supplier: Merck
UHPLC columns, InfinityLab Poroshell 120
Supplier: Agilent
D-(+)-Maltose monohydrate, Sigma-Aldrich®
Supplier: Merck