PHALLOIDIN-ATTO 488 BIOREAGENT, SUITABLE FOR FLUORESCENCE, >= 90% (HPCE) 1 * 1 Each
Catalog # SIAL49409-10NMOL
Supplier: Merck
PHALLOIDIN-ATTO 488 BIOREAGENT, SUITABLE FOR FLUORESCENCE, >= 90% (HPCE) 1 * 1 Each
Catalog # SIAL49409-10NMOL
Supplier: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Detergent, powder for automated cleaning of glassware, neodisher® LaboClean A8
Supplier: Dr. Weigert
HPLC column accessories
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection