Order Entry
Poland
Orders LinkContactUsLinkComponent
Nickel(II) carbonate basic hydrate, Sigma-Aldrich®

Nickel(II) carbonate basic hydrate, Sigma-Aldrich®

Plastic bottle for solids, 1 kg
Catalog # SIAL72248-1KG
Supplier:   Merck
Nickel(II) carbonate basic hydrate, Sigma-Aldrich®
Catalog # SIAL72248-1KG
Supplier:  Merck
CAS Number:  12607-70-4
Nickel(II) carbonate basic hydrate, Sigma-Aldrich®

Plastic bottle for solids, 1 kg

CAS Number:  12607-70-4
Price
573 zł
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Nickel(II) carbonate hydroxide hydrate, Basic nickel carbonate
  • CAS Number:12607-70-4
  • Formula:~NiCO₃ · 2Ni(OH)₂ · 4H₂O
  • MDL Number:MFCD00016262
  • Molecular Weight:304.11 g/mol (anhy)
  • EINECS:235-715-9

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.