Order Entry
Poland
Orders LinkContactUsLinkComponent
Empty columns, HiScale™
HiScale™ 26/40, Glass, 26×400 mm
Catalog # 28-9645-13
Supplier:   Cytiva
Empty columns, HiScale™
Catalog # 28-9645-13
Supplier:  Cytiva
Empty columns, HiScale™
Empty columns, HiScale™
HiScale™ 26/40, Glass, 26×400 mm
Price
7 040 zł
Log in for availability
1 * 1 SZT
Product Details & Documents
HiScale™ empty chromatography columns are a family of pressure-stable, easy to pack, columns for medium to high pressure liquid chromatography using a chromatography system.
  • Convenient column measurement scale, nonrotating plunger mechanism, and ergonomically designed components for easier adjustment during column packing
  • Built to withstand pressures up to 20 bar (2 MPa, 290 psi) for compatibility with modern BioProcess™ resins
  • Dual adapters and axial compression capability allow for a large range of bed heights and column packing protocols
  • Choose from 10, 16, 26, or 50 mm inner diameter, and 200 mm or 400 mm length to accommodate bed volumes up to 785 ml
  • Designed to facilitate robust process scaleup, as well as cost-efficient and accurate scaledown for applications such as viral clearance studies
  • Equipped with two adapters allowing for a large range of bed heights
HiScale™ pressure‐stable, empty columns are designed for process development and preparative chromatography using ÄKTA protein purification systems. These chromatography columns withstand pressures up to 20 bar, with a user‐friendly design that supports process optimization through simplified and reproducible operations.
The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments as well as disassembly and cleaning. The entire shaft may be rinsed after column packing, minimizing the risk of bacterial growth. The column’s measurement scale helps users to predict and reach a predefined bed height.
HiScale™ columns are based on the well‐established line of XK columns. They are compatible with all methods currently applicable to XK columns.
HiScale™ 10/40 columns are especially suitable for scaledown studies used in, for example, viral clearance evaluation in downstream processing. The small inner diameter and flexible adapters minimize costly sample and viral spike volumes. The robust design of HiScale™ 10/40 allows for controlled bed heights and gives reproducible results offering an effective scaled down model of the large-scale downstream process.
HiScale™ chromatography columns are compatible with all Cytiva resins. For columns packed with SOURCE 15 and SOURCE 30, a 10 µm net ring should be used.
Delivery information: Each HiScale™ column is delivered with a column tube, outer protection tube, two adapters, PEEK tubing, and unions at both ends for easy connection to valves, pumps, and monitors. For HiScale™ 10/40, connectors are provided in the accessory kit included in the package.
Specifications
  • Description:
    HiScale™ 26/40
  • Seq:
    05
  • Column hardware:
    Glass
  • Bed dimensions:
    26×400 mm
  • Cat. No.:
    28-9645-13
  • Max. pressure:
    20 bar
  • Temperature limit:
    4...40 °C
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Connector 1/16" male/Luer female, for HiScale™ 16 and 26
    Connector 1/16" male/Luer female, for HiScale™ 16 and 26
    18-1112-51
    Log in for availability
    1 * 2 SZT - 459 zł
Product Information
  • Catalog #
    Availability
    Price
  • Short column holder, for HiScale™ 16 and 26
    Short column holder, for HiScale™ 16 and 26
    18-1113-17
    Log in for availability
    1 * 1 SZT - 1 180 zł
Product Information
  • Catalog #
    Availability
    Price
  • Column clamp, OD 10 - 21 mm
    Column clamp, OD 10 - 21 mm
    28-9563-19
    Log in for availability
    1 * 5 SZT - 320 zł
Product Information
  • Catalog #
    Availability
    Price
  • Column holder, steel, for HiScale™ 50
    Column holder, steel, for HiScale™ 50
    28-9644-99
    Log in for availability
    1 * 1 SZT - 2 120 zł
Product Information
  • Catalog #
    Availability
    Price
  • Packing tube 40, for HiScale™ 26
    Packing tube 40, for HiScale™ 26
    28-9645-05
    Log in for availability
    1 * 1 SZT - 6 470 zł
Product Information
  • Catalog #
    Availability
    Price
  • Packing tube 40, for HiScale™ 50
    Packing tube 40, for HiScale™ 50
    28-9645-06
    Log in for availability
    1 * 1 SZT - 10 280 zł
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Spanner wrench, for nuts Ø 58 - 62 mm, for HiScale™ 16 columns
    Spanner wrench, for nuts Ø 58 - 62 mm, for HiScale™ 16 columns
    28-9647-76
    Galvanised steel
    Log in for availability
    1 * 2 SZT - 403 zł
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Spanner wrench, for nuts Ø 65 - 75 mm, for HiScale™ 26 columns
    Spanner wrench, for nuts Ø 65 - 75 mm, for HiScale™ 26 columns
    28-9647-77
    Galvanised steel
    Log in for availability
    1 * 2 SZT - 446 zł
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Spanner wrench, for nuts Ø 95 - 100 mm, for HiScale™ 50 columns
    Spanner wrench, for nuts Ø 95 - 100 mm, for HiScale™ 50 columns
    28-9647-78
    Galvanised steel
    Log in for availability
    1 * 2 SZT - 495 zł
Product Information
  • Catalog #
    Availability
    Price
  • Union Valco F/F, PEEK, for HiScale™ 16 and 26
    Union Valco F/F, PEEK, for HiScale™ 16 and 26
    11-0003-39
    Log in for availability
    1 * 5 SZT - 709 zł
Product Information
  • Catalog #
    Availability
    Price
  • Fingertight stop plug, 1/16", PEEK, for HiScale™ 16 and 26
    Fingertight stop plug, 1/16", PEEK, for HiScale™ 16 and 26
    11-0003-55
    Log in for availability
    1 * 5 SZT - 338 zł
Product Information
  • Catalog #
    Availability
    Price
  • Superloop, M6 fittings, 150 ml, for HiScale™ 16, 26 and 50
    Superloop, M6 fittings, 150 ml, for HiScale™ 16, 26 and 50
    18-1023-85
    Log in for availability
    1 * 1 SZT - 7 310 zł
Product Information
  • Catalog #
    Availability
    Price
  • Accessory kit for HiScale™ 16, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs
    Accessory kit for HiScale™ 16, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs
    28-9663-67
    Log in for availability
    1 * 1 KIT - 403 zł
Product Information
  • Catalog #
    Availability
    Price
  • Accessory kit for HiScale™ 26, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs
    Accessory kit for HiScale™ 26, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs
    28-9663-74
    Log in for availability
    1 * 1 KIT - 446 zł
Product Information
  • Catalog #
    Availability
    Price
  • Tubing, inner Ø 2 mm, for HiScale™ 50
    Tubing, inner Ø 2 mm, for HiScale™ 50
    28-9663-76
    Log in for availability
    1 * 1 m - 533 zł
Product Information
  • Catalog #
    Availability
    Price
  • Net ring, 20 μm, PP/PA (nylon), for HiScale™ 16
    Net ring, 20 μm, PP/PA (nylon), for HiScale™ 16
    28-9663-79
    Log in for availability
    1 * 5 SZT - 369 zł
Product Information
  • Catalog #
    Availability
    Price
  • Net ring, 20 μm, PP/PA (nylon), for HiScale™ 26
    Net ring, 20 μm, PP/PA (nylon), for HiScale™ 26
    28-9663-80
    Log in for availability
    1 * 5 SZT - 403 zł
Product Information
  • Catalog #
    Availability
    Price
  • Net ring, 20 μm, PP/PA (nylon), for HiScale™ 50
    Net ring, 20 μm, PP/PA (nylon), for HiScale™ 50
    28-9663-81
    Log in for availability
    1 * 5 SZT - 495 zł
Product Information
  • Catalog #
    Availability
    Price
  • Adapter, for HiScale™ 50
    Adapter, for HiScale™ 50
    28-9663-85
    Log in for availability
    1 * 1 SZT - 3 350 zł
Product Information
  • Catalog #
    Availability
    Price
  • Column tube 20, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 16
    Column tube 20, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 16
    28-9666-46
    Log in for availability
    1 * 1 SZT - 2 460 zł
Product Information
  • Catalog #
    Availability
    Price
  • Column tube 20, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 26
    Column tube 20, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 26
    28-9666-48
    Log in for availability
    1 * 1 SZT - 4 160 zł
Product Information
  • Catalog #
    Availability
    Price
  • Column tube 20, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 50
    Column tube 20, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 50
    28-9666-49
    Log in for availability
    1 * 1 SZT - 5 370 zł
Product Information
  • Catalog #
    Availability
    Price
  • Column tube 40, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 50
    Column tube 40, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 50
    28-9666-50
    Log in for availability
    1 * 1 SZT - 8 090 zł
Product Information
  • Catalog #
    Availability
    Price
  • Column tube 40, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 26
    Column tube 40, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 26
    28-9666-51
    Log in for availability
    1 * 1 SZT - 5 260 zł
Product Information
  • Catalog #
    Availability
    Price
  • Column tube 40, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 16
    Column tube 40, Borosilicate glass/PC/PA/EPDM/POM, for HiScale™ 16
    28-9666-52
    Log in for availability
    1 * 1 SZT - 3 190 zł
Product Information
  • Catalog #
    Availability
    Price
  • O-ring, packing tube, for HiScale™ 16
    O-ring, packing tube, for HiScale™ 16
    28-9666-53
    Log in for availability
    1 * 1 SZT - 548 zł
Product Information
  • Catalog #
    Availability
    Price
  • O-ring, 26×4 mm, for HiScale™ 26
    O-ring, 26×4 mm, for HiScale™ 26
    28-9666-54
    Log in for availability
    1 * 2 SZT - 441 zł
Product Information
  • Catalog #
    Availability
    Price
  • O-ring, 48×4 mm, for HiScale™ 50
    O-ring, 48×4 mm, for HiScale™ 50
    28-9666-55
    Log in for availability
    1 * 1 SZT - 853 zł
Product Information
  • Catalog #
    Availability
    Price
  • Tubing, outer Ø 2 mm, cut, for HiScale™ 50
    Tubing, outer Ø 2 mm, cut, for HiScale™ 50
    28-9666-56
    Log in for availability
    1 * 0,295 m - 403 zł
Product Information
  • Catalog #
    Availability
    Price
  • Packing tube 20, for HiScale™ 50
    Packing tube 20, for HiScale™ 50
    28-9802-51
    Log in for availability
    1 * 1 SZT - 7 150 zł
Product Information
  • Catalog #
    Availability
    Price
  • Packing tube 40, for HiScale™ 16
    Packing tube 40, for HiScale™ 16
    28-9868-15
    Log in for availability
    1 * 1 SZT - 5 100 zł
Product Information
  • Catalog #
    Availability
    Price
  • Packing tube 20, for HiScale™ 16
    Packing tube 20, for HiScale™ 16
    28-9868-16
    Log in for availability
    1 * 1 SZT - 3 900 zł
Product Information
  • Catalog #
    Availability
    Price
  • Packing tube 20, for HiScale™ 26
    Packing tube 20, for HiScale™ 26
    28-9803-83
    Log in for availability
    1 * 1 SZT - 5 340 zł
Product Information
  • Catalog #
    Availability
    Price
  • Accessory kit for HiScale™ 50, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs
    Accessory kit for HiScale™ 50, includes 1 net and O-ring, 1 support screen, 2 ferrules and 2 fingertight stop plugs
    28-9663-75
    Log in for availability
    1 * 1 KIT - 650 zł
Product Information
  • Catalog #
    Availability
    Price
  • Tubing, inner Ø 1 mm, for HiScale™ 16 and 26, beige
    Tubing, inner Ø 1 mm, for HiScale™ 16 and 26, beige
    18-1115-83
    Log in for availability
    1 * 1 SZT - 410 zł
Product Information
  • Catalog #
    Availability
    Price
  • Stop plug female, 1/16”
    Stop plug female, 1/16”
    11-0004-64
    Log in for availability
    1 * 5 SZT - 427 zł