Spoon spatulas
150 mm
Catalog # JUCHVL04010
Supplier: JUCHHEIM Laborgerate GmbH
Jump to:
Product Details & Documents
Double ended spoon spatulas made from stainless steel.
- One broad spatula end, one spoon end
Specifications
- Length:150 mm
- Seq:01