Order Entry
Poland
Orders LinkContactUsLinkComponent
Aluminium oxide basic for TLC

Aluminium oxide basic for TLC

500 g
Catalog # 46001.36
Aluminium oxide basic for TLC
Catalog # 46001.36
CAS Number:  1344-28-1
Aluminium oxide basic for TLC
Aluminium oxide basic for TLC

500 g

CAS Number:  1344-28-1
Price
1 020 zł
Log in for availability
1 * 500 g
Product Details & Documents

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 5-15 Micron APS Powder Refractive Index: 1.765 Appearance: White to pale pink Odour: Odourless Melting Point: 2045° Boiling Point: 2980° MDL No.: MFCD00003424

  • CAS Number:1344-28-1
  • Formula:Al₂O₃
  • MDL Number:MFCD00003424
  • Molecular Weight:101.96 g/mol
  • Merck Index:14,356
  • Boiling Pt:2980 °C
  • Melting Pt:2045 °C
  • Storage Temperature:Ambient
  • EINECS:215-691-6
Specifications

Specifications

  • Pk:
    500 g

Specifications