Aluminium oxide basic for TLC
100 g
Supplier: Thermo Fisher Scientific
Aluminium oxide basic for TLC
Catalog # 46001.22
Supplier: Thermo Fisher Scientific
CAS Number: 1344-28-1
Jump to:
Product Details & Documents
CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 5-15 Micron APS Powder Refractive Index: 1.765 Appearance: White to pale pink Odour: Odourless Melting Point: 2045° Boiling Point: 2980° MDL No.: MFCD00003424
- CAS Number:1344-28-1
- Formula:Al₂O₃
- MDL Number:MFCD00003424
- Molecular Weight:101.96 g/mol
- Merck Index:14,356
- Boiling Pt:2980 °C
- Melting Pt:2045 °C
- Storage Temperature:Ambient
- EINECS:215-691-6
Documents
Specifications
Specifications
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



