ADENOSINE 5'-MONOPHOSPHATE SODIUMFROM YE AST 1 * 5 g
SIALA1752-5G
: Merck
Frequently Bought Together
2-PROPANOL 70% R-R ATOMIZER 1 * 1 L
: CHEMPUR
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
2-Propanol, pure
: CHEMPUR
N,N-Dimethylacetamide 99%, pure
: Thermo Fisher Scientific
Toluene, pure analytical reagent
: POCH AVANTOR BRAND
Bis(2-methoxyethyl)ether, extra dry 99+%, AcroSeal®
: Thermo Fisher Scientific
4-Methyl-2-pentanone 98.5+% ACS
: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabiliser free, AcroSeal®
: Thermo Fisher Scientific
2-Propanol ≥99.7% for gas chromatography
: Thermo Fisher Scientific
Butyl acetate 99+%, extra pure
: Thermo Fisher Scientific