Order Entry
Poland
Orders LinkContactUsLinkComponent
NMR STD/ACETONE-D6, 100% (99.96 ATOM %& 1 * 1 Each

NMR STD/ACETONE-D6, 100% (99.96 ATOM %& 1 * 1 Each

Catalog # SIAL708941-7IN
Supplier:   Merck
NMR STD/ACETONE-D6, 100% (99.96 ATOM %& 1 * 1 Each
Catalog # SIAL708941-7IN
Supplier:  Merck
CAS Number:  
NMR STD/ACETONE-D6, 100% (99.96 ATOM %& 1 * 1 Each
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 Szt.
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.