Citrate-phosphate-dextrose solution with adenine 10x50ML 1 * 10 items
SIALC4431-10X50ML
: Merck
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
epT.I.P.S.® Bulk - Pipette Tips
: EPPENDORF
Water, TECHNICAL, demineralised
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals