DISCOVERY DSC-18 PK/30 6ML TUBE 1 GRAM 1 * 30 items
Catalog # 52606-U
Supplier: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Florisil® 60-100 mesh for chromatography
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Exhaust filters, V3.0
Supplier: S.C.A.T.
Folin-Ciocalteu's reagent for analysis of phenols, Supelco®
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals