Order Entry
Poland
Orders LinkContactUsLinkComponent
Gastro-norm container
6,5 l, 325×265×100 mm
  HUPF0130782H
Gastro-norm container
  HUPF0130782H
Gastro-norm container
Gastro-norm container
6,5 l, 325×265×100 mm
351 zł
1 * 1 SZT
Stainless steel 18/10.
  • Fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance
Multipurpose containers for a wide range of uses.
  • Capacity:
    6,5 l
  • Seq:
    01
  • W×D×H ext.:
    325×265×100 mm