0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
FREEZING POINT STD CRYOSCOPE 530 100ML 1 * 1 items
REGEMTR030X
:
REAGECON
FREEZING POINT STD CRYOSCOPE 530 100ML 1 * 1 items
REGEMTR030X
:
REAGECON
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
VWR®, Chlorate 1,000 mg/L in Water standard for ion chromatography
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Phosphorus standard solution, 1,000 mg/L P in water AVS TITRINORM standard for AAS
: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
: Merck
pH buffers, ready to use
: WTW