Order Entry
Poland
ContactUsLinkComponent
Tubing, PEEK, 1/32 x 50 um / .002 ID, orange solid, 3 m/pkg 1 * 3 m
Catalog # JOURJR-T-5602-M3
Supplier:  VICI JOUR RESEARCH
Tubing, PEEK, 1/32 x 50 um / .002 ID, orange solid, 3 m/pkg 1 * 3 m
Catalog # JOURJR-T-5602-M3
Supplier:  VICI JOUR RESEARCH
Frequently Bought Together