OXALATE IC-STANDARD 1 * 100 mL
Catalog # REGEICAS13
Supplier: REAGECON
Frequently Bought Together
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR® ARISTAR®, Cadmium Standard Solutions for ICP-MS
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
Conductivity standards
Supplier: HAMILTON BONADUZ
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Silicone Rotational viscosity standards
Supplier: VWR Chemicals
Malachite green solution Reag. Ph. Eur. 1050501
Supplier: VWR Chemicals
Turbidity standards, Stablcal®
Supplier: Hach
pH buffer solutions, colour coded
Supplier: Hach
Pipette tips, ART®, SoftFit L™
Supplier: Thermo Fisher Scientific
VWR®, Cold Cranking Simulator (CCS) viscosity standards
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond