Order Entry
Poland
Orders LinkContactUsLinkComponent
BUFFER PH 1.679 25°C IUPAC + CERTIF.

BUFFER PH 1.679 25°C IUPAC + CERTIF.

Catalog # 665-0394
Supplier:   Hach
BUFFER PH 1.679 25°C IUPAC + CERTIF.
Catalog # 665-0394
Supplier:  Hach
BUFFER PH 1.679 25°C IUPAC + CERTIF.
BUFFER PH 1.679 25°C IUPAC + CERTIF.
Price
338 zł
Log in for availability
1 * 500 mL
Product Details & Documents