(2-Hydroxylpropyl)-β-cyclodextrine
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
(2-Hydroxylpropyl)-β-cyclodextrine
Catalog # H31133.14
Supplier: Thermo Fisher Scientific
CAS Number: 128446-35-5
Jump to:
Product Details & Documents
CAS No.: 128446-35-5 Melting Point: 305° MDL No.: MFCD00069372
Synonyms: Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD
- CAS Number:128446-35-5
- Formula:C₆₃H₁₁₂O₄₂
- MDL Number:MFCD16621721
- Melting Pt:305 °C
- Storage Temperature:Ambient
- EINECS:420-920-1
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



