Order Entry
Poland
Orders LinkContactUsLinkComponent
(2-Hydroxylpropyl)-β-cyclodextrine

(2-Hydroxylpropyl)-β-cyclodextrine

Glass bottle for solids, 5 g
Catalog # H31133.06
(2-Hydroxylpropyl)-β-cyclodextrine
Catalog # H31133.06
CAS Number:  128446-35-5
(2-Hydroxylpropyl)-β-cyclodextrine
(2-Hydroxylpropyl)-β-cyclodextrine

Glass bottle for solids, 5 g

CAS Number:  128446-35-5
Price
365 zł
Log in for availability
1 * 5 g
Product Details & Documents

CAS No.: 128446-35-5 Melting Point: 305° MDL No.: MFCD00069372

Synonyms:  Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD
  • CAS Number:128446-35-5
  • Formula:C₆₃H₁₁₂O₄₂
  • MDL Number:MFCD16621721
  • Melting Pt:305 °C
  • Storage Temperature:Ambient
  • EINECS:420-920-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications