Order Entry
Poland
ContactUsLinkComponent
Flask volumetric, capacity 20ml, poly stopper, class A, NS 12/21, amber graduation, conformity certified. 1 * 2 items
Catalog # HIRS2640166
Supplier:  HIRSCHMANN
Flask volumetric, capacity 20ml, poly stopper, class A, NS 12/21, amber graduation, conformity certified. 1 * 2 items
Catalog # HIRS2640166
Supplier:  HIRSCHMANN
Frequently Bought Together