Order Entry
Poland
Orders LinkContactUsLinkComponent
Pipette controller, pipetus®-standard
pipetus®-standard complete, UK-plug
  HIRS9903010
Pipette controller, pipetus®-standard
  HIRS9903010
 :  
Pipette controller, pipetus®-standard
Pipette controller, pipetus®-standard
pipetus®-standard complete, UK-plug
Pipette controller for pipettes from 0,1 to 100 ml.
  • Optimum safety and excellent convenience of use
  • Pipettor is lighter and easy to use because the pump is separated from the handle
  • Autoclavable, detachable handle for the pipette holder is equipped with two buttons for filling and emptying
This electric pipettor is equipped with a 0,2 µm integrated water-repellent filter (for better durability) and a safety valve to protect the equipment from the introduction of hazardous (contaminated or radioactive) liquids.
 : The complete standard pipetus® is supplied with an electric pump, two filters and a safety valve.
  • Description:
    pipetus®-standard complete, UK-plug
  • Seq:
    02
Accessories
  •  
    Description
  • HIRS9904400
    External pump with filter, 230 V, EU
    1 * 1 SZT - 876 zł
  •  
    Description
  • 612-2615
    Silicone adapter
    1 * 1 SZT - 41,70 zł
  •  
    Description
  • HIRS9903705
    Round filter set with one-sided connection (50 filters)
    1 * 50 SZT - 1 480 zł
  •  
    Description
  • 612-2616
    Safety valve
    1 * 1 SZT - 155 zł
  •  
    Description
  • 612-2618
    Microfibre-filter set for pump (5 filters)
    1 * 5 SZT - 3 400 zł
  •  
    Description
  • HIRS9903205
    Spare O-rings for pipetus® akku and pipetus®
    1 * 5 SZT - 201 zł
  •  
    Description
  • HIRS9903300
    Pipette holder, complete, turquoise, for pipetus®/pipetus® accu/pipetus® standard/pipetus® classic
    1 * 1 SZT - 295 zł
  •  
    Description
  • 612-1982
    Round filter set with one-sided connection (5 filters)
    1 * 5 SZT - 170 zł
  •  
    Description
  • HIRS9903714
    Filter set, sterile, individually packed, for pipetus®
    1 * 5 SZT - 1 070 zł
  •  
    Description
  • HIRS9903706
    Round filter set with one-sided connection (100 filters)
    1 * 100 SZT - 2 560 zł