Pipette controller, pipetus®-standard
pipetus®-standard complete with extra filter on pump, UK-plug
Catalog # HIRS9904010
Supplier: HIRSCHMANN
Przejdź do:
Product Details & Documents
Pipette controller for pipettes from 0,1 to 100 ml.
- Optimum safety and excellent convenience of use
- Pipettor is lighter and easy to use because the pump is separated from the handle
- Autoclavable, detachable handle for the pipette holder is equipped with two buttons for filling and emptying
This electric pipettor is equipped with a 0,2 µm integrated water-repellent filter (for better durability) and a safety valve to protect the equipment from the introduction of hazardous (contaminated or radioactive) liquids.
Ordering information: The complete standard pipetus® is supplied with an electric pump, two filters and a safety valve.
Documents
Hirschmann pipetus® HiClass in pipetting
Product Literature
pipetus® standard
Images 360
Dane techniczne
- Description:pipetus®-standard complete with extra filter on pump, UK-plug
- Seq:04
Accessories
Informacje o produkcie
- Catalog #AvailabilityPrice
- External pump with filter, 230 V, EU
Informacje o produkcie
- Catalog #AvailabilityPrice
- Silicone adapter
Informacje o produkcie
- Catalog #AvailabilityPrice
- Round filter set with one-sided connection (50 filters)Round filter set with one-sided connection (50 filters)HIRS9903705Zaloguj się dla dostępności1 * 50 SZT - 1 480 zł
Informacje o produkcie
- Catalog #AvailabilityPrice
- Safety valve
Informacje o produkcie
- Catalog #AvailabilityPrice
- Microfibre-filter set for pump (5 filters)Microfibre-filter set for pump (5 filters)612-2618Zaloguj się dla dostępności1 * 5 SZT - 3 400 zł
Informacje o produkcie
- Catalog #AvailabilityPrice
- Spare O-rings for pipetus® akku and pipetus®Spare O-rings for pipetus® akku and pipetus®HIRS9903205Zaloguj się dla dostępności1 * 5 SZT - 201 zł
Informacje o produkcie
- Catalog #AvailabilityPrice
- Pipette holder, complete, turquoise, for pipetus®/pipetus® accu/pipetus® standard/pipetus® classicPipette holder, complete, turquoise, for pipetus®/pipetus® accu/pipetus® standard/pipetus® classicHIRS9903300Zaloguj się dla dostępności1 * 1 SZT - 295 zł
Informacje o produkcie
- Catalog #AvailabilityPrice
- Round filter set with one-sided connection (5 filters)Round filter set with one-sided connection (5 filters)612-1982Zaloguj się dla dostępności1 * 5 SZT - 170 zł
Informacje o produkcie
- Catalog #AvailabilityPrice
- Filter set, sterile, individually packed, for pipetus®Filter set, sterile, individually packed, for pipetus®HIRS9903714Zaloguj się dla dostępności1 * 5 SZT - 1 070 zł
Informacje o produkcie
- Catalog #AvailabilityPrice
- Round filter set with one-sided connection (100 filters)Round filter set with one-sided connection (100 filters)HIRS9903706Zaloguj się dla dostępności1 * 100 SZT - 2 560 zł
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



