Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TRYPSIN SOLUTION FROM FROM PORCINE PANCREAS 10×, BIOREAGENT, 25 G PORCINE TRYPSIN PER LITER IN HANKS' BALANCED SALT SOL 1 * 100 mL
Catalog # SIALT4674-100ML
Supplier:
Merck
Expand image
TRYPSIN SOLUTION FROM FROM PORCINE PANCREAS 10×, BIOREAGENT, 25 G PORCINE TRYPSIN PER LITER IN HANKS' BALANCED SALT SOL 1 * 100 mL
Catalog # SIALT4674-100ML
Supplier:
Merck
Frequently Bought Together
Dry heat indicator tapes
Supplier: KARTELL
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Multiwell Cell Culture Plates
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Supelco® PESTANAL® Organic Reference Standard, Glyphosate
Supplier: Merck
MINIMUM ESSENTIAL MEDIUM EAGLE, WITH EARLE'S SALTS AND L-GLUTAMINE, WITHOUT SODIUM BICARBONATE. CELL CULTURE TESTED 1 * 10 L
Supplier: Merck