1,3,5,7,9,11,14-HEPTA-ISOOCTYLTRICYCLO(7.) HEPTASILOXANE-ENDO-3,7,14-TRIOL, 95% 1 * 5 g
Catalog # SIAL560391-5G
Supplier: Merck
1,3,5,7,9,11,14-HEPTA-ISOOCTYLTRICYCLO(7.) HEPTASILOXANE-ENDO-3,7,14-TRIOL, 95% 1 * 5 g
Catalog # SIAL560391-5G
Supplier: Merck
Frequently Bought Together
Methyl ethyl ketone ≥99%, TECHNICAL
Supplier: VWR Chemicals
4-tert-Butylbenzoic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
2-Propanol 70 % pure 1 * 1 L
Supplier: PanReac AppliChem