QUERCETIN ANHYDROUS 1 * 10 g
SIALQ4951-10G
: Merck
Frequently Bought Together
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
: MERCK PRODUCTION CHEMICALS
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals