1,3,5,7,9,11,13,15-OCTAPHENYLPENTACYCLO-OCTASILOXANE 1 * 25 g
Catalog # SIAL526851-25G
Supplier: Merck
1,3,5,7,9,11,13,15-OCTAPHENYLPENTACYCLO-OCTASILOXANE 1 * 25 g
Catalog # SIAL526851-25G
Supplier: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Carbon disulphide ≥99.9%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS for analysis
Supplier: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals