L-ALPHA-PHOSPHATIDYLCHOLINE FROM DRIED EGG YOLK, TYPE X-E 1 * 10 g
Catalog # SIALP5394-10G
Supplier: Merck
L-ALPHA-PHOSPHATIDYLCHOLINE FROM DRIED EGG YOLK, TYPE X-E 1 * 10 g
Catalog # SIALP5394-10G
Supplier: Merck
Frequently Bought Together
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
tert-Butyl methyl ether, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Cyclohexanone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Benzoic acid methyl ester
Supplier: Merck
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
2-PROPANOL CZDA OPAK. Z TW. 1 * 5 L
Supplier: CHEMPUR
, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals