MEDIUM 199, 10×, WITH HANKS' SALTS, WITHOUT L-GLUTAMINE AND SODIUM BICARBONATE, STERILE-FILTERED. CELL CULTURE TESTED 1 * 500 mL
SIALM9163-500ML
: Merck
MEDIUM 199, 10×, WITH HANKS' SALTS, WITHOUT L-GLUTAMINE AND SODIUM BICARBONATE, STERILE-FILTERED. CELL CULTURE TESTED 1 * 500 mL
SIALM9163-500ML
: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection