CARBON NANOTUBE, MULTI-WALLED, AS-PRODUCED CATHODE DEPOSIT, >7.5% MWCNT BASIS, O.D. X L 7-15 NM X 0.5-10 UM 1 * 2 g
SIAL412988-2G
: Merck
CARBON NANOTUBE, MULTI-WALLED, AS-PRODUCED CATHODE DEPOSIT, >7.5% MWCNT BASIS, O.D. X L 7-15 NM X 0.5-10 UM 1 * 2 g
SIAL412988-2G
: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection