4,4'-METHYLENEBIS(CYCLOHEXYLAMINE), TECH ., 95%, MIXTURE OF ISOMERS 1 * 250 g
Catalog # SIAL368849-250G
Supplier: Merck
4,4'-METHYLENEBIS(CYCLOHEXYLAMINE), TECH ., 95%, MIXTURE OF ISOMERS 1 * 250 g
Catalog # SIAL368849-250G
Supplier: Merck
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Petroleum spirit, 40…60 °C, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
1-Propanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine (synthetic) 99+%, pure
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dilithium oxide ≥99.5% (metals basis)
Supplier: Thermo Fisher Scientific