BETA-ALANINE ETHYL ESTER HYDROCHLORIDE, 98% 1 * 10 g
Catalog # SIAL306142-10G
Supplier: Merck
Frequently Bought Together
Ethyl acetate, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Acetone, pure
Supplier: CHEMPUR
Dichloromethane stabilized with 20 ppm of amylene for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Acetone, for GC Plus, Burdick & Jackson™
Supplier: Honeywell Chemicals
Water, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
Cesium fluoride 99.9%, extra pure
Supplier: Thermo Fisher Scientific
Dichloromethane, anhydrous ≥99.7% stabilised
Supplier: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Acetone (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 1 L
Supplier: PanReac AppliChem
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
EX-CELL Advanced CHO Fed-batch Medium 1 * 1.000 mL
Supplier: Merck
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
o-Xylene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanolamine, Sigma-Aldrich®
Supplier: Merck