Order Entry
Poland
Orders LinkContactUsLinkComponent
Nickel, Sigma-Aldrich®

Nickel, Sigma-Aldrich®

Glass bottle for solids, 1,4 g
Catalog # SIAL267007-1.4G
Health
Supplier:   Merck
Nickel, Sigma-Aldrich®
Catalog # SIAL267007-1.4G
Supplier:  Merck
CAS Number:  7440-02-0
Nickel, Sigma-Aldrich®

Glass bottle for solids, 1,4 g

CAS Number:  7440-02-0
Price
1 080 zł
Log in for availability
1 * 1,4 g
Product Details & Documents
  • CAS Number:7440-02-0
  • Formula:Ni
  • MDL Number:MFCD00011137
  • Molecular Weight:58.69 g/mol
  • Merck Index:12,06582
  • Boiling Pt:3007 °C (1013 hPa)
  • Melting Pt:1453 °C
  • Density:8.9 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-111-4
  • ADR:4.1,III
  •  :M-Clarity™ quality level = MQ100
  • UN:3089
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1,4 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.