Order Entry
Poland
Orders LinkContactUsLinkComponent
Methyl-4-hydroxybenzoate, Sigma-Aldrich®

Methyl-4-hydroxybenzoate, Sigma-Aldrich®

1 Vial
Catalog # SIALM8911-1VL
Irritant
Supplier:   Merck
Methyl-4-hydroxybenzoate, Sigma-Aldrich®
Catalog # SIALM8911-1VL
Supplier:  Merck
CAS Number:  99-76-3
Methyl-4-hydroxybenzoate, Sigma-Aldrich®
Methyl-4-hydroxybenzoate, Sigma-Aldrich®

1 Vial

CAS Number:  99-76-3
Price
798 zł
Log in for availability
1 * 1 Vial
Product Details & Documents
Synonyms:  Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
  • CAS Number:99-76-3
  • Formula:HOC₆H₄CO₂CH₃
  • MDL Number:MFCD00002352
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,06129
  • Boiling Pt:270…280 °C (1013 hPa)
  • Melting Pt:125…128 °C
  • Density:1.1208 g/cm³ (137 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:202-785-7
Specifications

Specifications

  • Pk:
    1 Vial

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.