Order Entry
Poland
Orders LinkContactUsLinkComponent
Methyl-4-hydroxybenzoate, Sigma-Aldrich®
5 Vial
  SIALM8911-5VL
 :   Merck
Methyl-4-hydroxybenzoate, Sigma-Aldrich®
  SIALM8911-5VL
 :  Merck
 :  99-76-3
Methyl-4-hydroxybenzoate, Sigma-Aldrich®
Methyl-4-hydroxybenzoate, Sigma-Aldrich®
5 Vial
 :  99-76-3
3 180 zł
1 * 5 Vial
 :  Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
  •  :99-76-3
  •  :HOC₆H₄CO₂CH₃
  •  :MFCD00002352
  •  :152.15 g/mol
  •  :13,06129
  •  :270…280 °C (1013 hPa)
  •  :125…128 °C
  •  :1.1208 g/cm³ (137 °C)
  •  :M-Clarity™ quality level = MQ200
  •  :Ambient
  •  :202-785-7
  • Pk:
    5 Vial