α-Cyclodextrin, Sigma-Aldrich®
Glass bottle for solids, 1 g
Catalog # SIALC4642-1G
Supplier: Merck
Jump to:
Product Details & Documents
- CAS Number:10016-20-3
- Formula:C₃₆H₆₀O₃₀
- MDL Number:MFCD00078207
- Molecular Weight:972.84 g/mol
- :M-Clarity™ quality level = MQ300
- EINECS:233-007-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



