PROPYLMAGNESIUM CHLORIDE, 2.0M SOLUTION IN DIETHYL ETHER 1 * 800 mL
Catalog # SIAL224391-800ML
Supplier: Merck
PROPYLMAGNESIUM CHLORIDE, 2.0M SOLUTION IN DIETHYL ETHER 1 * 800 mL
Catalog # SIAL224391-800ML
Supplier: Merck
Frequently Bought Together
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Diethyl ether stabilised, EMPLURA®, Supelco®
Supplier: Merck
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Glass snap top vials, 2 ml, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals