Order Entry
Poland
Orders LinkContactUsLinkComponent
Selenium, Sigma-Aldrich®

Selenium, Sigma-Aldrich®

Glass bottle for solids, 50 g
Catalog # SIAL209643-50G
Toxic
Health
Supplier:   Merck
Selenium, Sigma-Aldrich®
Catalog # SIAL209643-50G
Supplier:  Merck
CAS Number:  7782-49-2
Selenium, Sigma-Aldrich®

Glass bottle for solids, 50 g

CAS Number:  7782-49-2
Price
672 zł
Log in for availability
1 * 50 g
Product Details & Documents
  • CAS Number:7782-49-2
  • Formula:Se
  • MDL Number:MFCD00134090
  • Molecular Weight:78.96 g/mol
  • Merck Index:12,08572
  • Boiling Pt:∼685 °C (1013 hPa)
  • Melting Pt:217 °C
  • Density:4.28 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-957-4
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ100
  • UN:3283
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.